Learn More
Hach Company Aluminum Reagent Set, Aluminon
Supplier:  Hach Company 2242000
Encompass
Reagent set for determination of aluminum by the AluVer3 Aluminon method. Hach Method 8012. Range: 0.008 - 0.800 mg/L Al. Set contains 100 each: AluVer3 Aluminon Reagent Powder Pillows, Ascorbic Acid Powder Pillows and Bleaching 3 Reagent Powder Pillows. 100 tests.
Catalog No. 50-199-5607