Learn More
Hach Company Total Nitrogen Reagent Set, HR, TNT
Supplier:  Hach Company 2714100
Encompass
For determination of Total Nitrogen by the Persulfate Digestion Test 'N Tube method: Hach Method 10072. Range: 2 - 150 mg/L N. 25-50 tests (dependent upon the number of reagent blank tests performed)
Catalog No. 50-199-6590