Learn More
Hach Company Total Nitrogen Reagent Set, LR, TNT
Supplier:  Hach Company 2672245
Encompass
For determination of Total Nitrogen by the Persulfate Digestion Test 'N Tube method: Hach Method 10071. Range: 0.5 - 25.0 mg/L N. 25-50 tests
Catalog No. 50-199-6519