Learn More
Hach Company Total Inorganic Nitrogen Pretreatment Reagent Set
Supplier:  Hach Company 2604945
Encompass
Pretreatment Reagent Set for determination of Total Inorganic Nitrogen by the Titanium Trichloride Reduction Test 'N Tube™ Method. Hach Method 10021. Used with Nitrogen-Ammonia Reagent Set, TNT, AmVer (Salicylate), LR (2604545). 25-50 tests (dependent upon the number of reagent blank tests performed).
Catalog No. 50-199-6326