Learn More
Hach Company Hardness (ULR) Reagent Set, Solution Pillows
Supplier:  Hach Company 2603100
Encompass
Reagent set ultra low range hardness determination by the Chlorophosphonazo method. Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3. Chlorophosphonazo reagent solution is packaged in unit-dose solution pillows. (100 tests).
Catalog No. 50-199-6312