Learn More
Hach Company Zinc Reagent Set, Zincon, 50 mL
Supplier:  Hach Company 2244800
Encompass
For determination of Zinc by the ZincoVer5 Zincon Method. USEPA approved for wastewater analysis (digestion is required for total zinc determination) using Hach Method 8009. Range: 0.01 - 3.00 mg/L Zn. Sample Size: 50 mL Set contains Cyclohexanone and ZincoVer 5 Powder Pillows for 100 tests. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5632