Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rgw-611 | 6497-78-5 | 98.0% | 226.24 | C9H14N4O3 | 10 MG
SDP

Catalog No. 5000240279
Encompass_Preferred

RGW-611 is a morpholine-derived research compound reported to enhance radiation-induced cell death in hypoxic V79-379A cells and to stimulate fatty acid synthesis. The compound is intended for laboratory research use and includes formulation, solubility, and storage guidance to support in vitro and in vivo preparation.

  • Enhances radiation-induced cell death in hypoxic V79-379A cells.
  • Stimulates fatty acid synthesis.
  • Chemical formula C9H14N4O3; molecular weight 226.24.
  • High DMSO solubility: 125 mg/mL with ultrasonic/warming to 60°C.
  • In vivo vehicle solubility ≥2.08 mg/mL reported for multiple formulations.
  • Storage: 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes with stock solution preparation volumes provided.

Catalog No. 50-002-40279 Supplier Medchemexpress LLC Supplier No. HY10784010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

RGW-611 is a morpholine-derived research compound reported to enhance radiation-induced cell death in hypoxic V79-379A cells and to stimulate fatty acid synthesis. The compound is intended for laboratory research use and includes formulation, solubility, and storage guidance to support in vitro and in vivo preparation.

  • Enhances radiation-induced cell death in hypoxic V79-379A cells.
  • Stimulates fatty acid synthesis.
  • Chemical formula C9H14N4O3; molecular weight 226.24.
  • High DMSO solubility: 125 mg/mL with ultrasonic/warming to 60°C.
  • In vivo vehicle solubility ≥2.08 mg/mL reported for multiple formulations.
  • Storage: 4°C protected from light; in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small research pack sizes with stock solution preparation volumes provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.