Learn More
Medchemexpress LLC Rgx-104 hydrochloride | 610318-03-1 | 99.9% | 632.54 | 1 MG
SDP
Catalog No. 5000345581
Encompass_Preferred
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. It significantly suppresses the growth of multiple cancer types in animals, including lung cancer, melanoma, glioblastoma, ovarian, renal cell, triple-negative breast, and colon cancer, with some instances showing partial or complete tumor regression.
  • Small-molecule LXR agonist.
  • Modulates innate immunity via transcriptional activation of the ApoE gene.
  • Significantly suppresses the growth of multiple cancer types in animals.
  • Treatment causes partial or complete tumor regression in some instances.
  • Available in solid form and in solution.
Catalog No. 50-003-45581 Supplier Medchemexpress LLC Supplier No. HY11149810MG
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. It significantly suppresses the growth of multiple cancer types in animals, including lung cancer, melanoma, glioblastoma, ovarian, renal cell, triple-negative breast, and colon cancer, with some instances showing partial or complete tumor regression.
  • Small-molecule LXR agonist.
  • Modulates innate immunity via transcriptional activation of the ApoE gene.
  • Significantly suppresses the growth of multiple cancer types in animals.
  • Treatment causes partial or complete tumor regression in some instances.
  • Available in solid form and in solution.