Learn More
Medchemexpress LLC RGX-104 (hydrochloride) | 610318-03-1 | 99.9% | 632.54 | 10 MG
SDP
Catalog No. 5000345690
Encompass_Preferred
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. It is for research use only and not sold to patients.
  • LXR agonist
  • Modulates innate immunity via transcriptional activation of the ApoE gene
  • Oral administration significantly suppresses the growth of multiple cancer types (e.g., lung cancer, melanoma, glioblastoma, ovarian, renal cell, triple-negative breast, and colon cancer) in animals with palpable tumors.
  • Can cause partial or complete tumor regression in some instances.
Catalog No. 50-003-45690 Supplier Medchemexpress LLC Supplier No. HY11149850MG
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. It is for research use only and not sold to patients.
  • LXR agonist
  • Modulates innate immunity via transcriptional activation of the ApoE gene
  • Oral administration significantly suppresses the growth of multiple cancer types (e.g., lung cancer, melanoma, glioblastoma, ovarian, renal cell, triple-negative breast, and colon cancer) in animals with palpable tumors.
  • Can cause partial or complete tumor regression in some instances.