Learn More
Medchemexpress LLC RGX-104 hydrochloride | 610318-03-1 | 99.9% | 632.54 | 25 MG
SDP
Catalog No. 5000346144
Encompass_Preferred
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. This compound is for research use only and not sold to patients. Studies show it significantly suppresses the growth of multiple cancer types in animals, even in large tumors, with instances of partial or complete tumor regression.
  • Acts as a small-molecule LXR agonist
  • Modulates innate immunity through ApoE gene activation
  • Significantly suppresses various cancer types in animal models
  • Effective against large tumors, with potential for regression
  • Applicable across a wide spectrum of malignancies
Catalog No. 50-003-46144 Supplier Medchemexpress LLC Supplier No. 25MGHY111498
RGX-104 hydrochloride is a small-molecule LXR agonist that modulates innate immunity via transcriptional activation of the ApoE gene. This compound is for research use only and not sold to patients. Studies show it significantly suppresses the growth of multiple cancer types in animals, even in large tumors, with instances of partial or complete tumor regression.
  • Acts as a small-molecule LXR agonist
  • Modulates innate immunity through ApoE gene activation
  • Significantly suppresses various cancer types in animal models
  • Effective against large tumors, with potential for regression
  • Applicable across a wide spectrum of malignancies