Learn More
TARGETMOL CHEMICALS INC RGX-202 500MG
SDP
Catalog No. 502576308
Encompass_Preferred
Also available in 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. RGX-202 ((beta)-GPA) is a creatine analog that alters skeletal muscle energy expenditure. It reduces cellular ATP creatine and phosphocreatine levels and stimulates AMP-activated protein kinase (AMPK) activating PPAR(gamma) coactivator 1(alpha) (PGC-1(alpha)). purity: 98%
Catalog No. 50-257-6308 Supplier TARGETMOL CHEMICALS INC Supplier No. T7535500MG
Also available in 50 mg 100 mg 200 mg and bulk. Please contact Fisher for quotes. RGX-202 ((beta)-GPA) is a creatine analog that alters skeletal muscle energy expenditure. It reduces cellular ATP creatine and phosphocreatine levels and stimulates AMP-activated protein kinase (AMPK) activating PPAR(gamma) coactivator 1(alpha) (PGC-1(alpha)). purity: 98%