Learn More
Medchemexpress LLC HY-W015828 50mg , RGX-202 CAS:353-09-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW01582850MG
Encompass_Preferred
HY-W015828 50mg RGX-202 CAS: 353-09-3 RGX-202 (3-Guanidinopropionic acid) is an oral small-molecule SLC6A8 transporter inhibitor. RGX-202 robustly inhibits creatine import in vitro and in vivo, reduces intracellular phosphocreatine and ATP levels, and induces tumor apoptosis. RGX-202 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1599