Learn More
Abcam Recombinant Human Histone H1 protein, 100UG
Supplier:  Abcam AB198676100UG
Encompass_Preferred
Recombinant Human Histone H1 protein is a Human Full Length protein, in the 2 to 194 aa range, expressed in Escherichia coli, with >=90% purity and suitable for SDS-PAGE.
Protein/ Peptide/ Enzyme Synonyms: H1F0, H1FV, H1-0, Histone H1.0, Histone H1', Histone H1(0)
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1690851