Learn More
Cole-Parmer Traceable Relative Humidity Meter with Dew Point and Calibration
Catalog No. NC1472918 Shop All Cole-Parmer Products
Encompass_Preferred
Pocket sized meter covers entire RH rangefrom 0 to 100%! Includes NIST-Traceable certificate from A2LA accredited (1750.01) calibration laboratory. Full-range response time is less than 1 minute. Sensor is not affected by condensation; measure up to 100% RH.
Catalog No. NC1472918 Supplier Cole-Parmer Supplier No. 9876688
Pocket sized meter covers entire RH rangefrom 0 to 100%! Includes NIST-Traceable certificate from A2LA accredited (1750.01) calibration laboratory. Full-range response time is less than 1 minute. Sensor is not affected by condensation; measure up to 100% RH.