Learn More
Enzo Life Sciences PDI (human), (recombinant) (His-tag) (1 mg)
SDP
Supplier:  Enzo Life Sciences ADISPP891J
Encompass_Preferred
Alternative name: P4HB, protein disulfide isomerase. MW: ~58kDa. Purity: ≥95% (SDS-PAGE; Western blot). Formulation: Liquid. In 50mM TRIS, pH 7.5, containing 150mM sodium chloride, 1mM DTT, 1mM EDTA, and 5% glycerol. Source: Produced in E. coli. Human PDI is fused at the N-terminus to a His-tag. UniProt: P07237. Application: WB, Activity assay. Long Term Storage: -80°C
Catalog No. NC1722585