Learn More
Bellco Glass, Inc. RH Pitch Blade Imp 2 3/4in,7L
SDP
Catalog No. 501946442
Encompass_Preferred
Sst pitched blade impeller with right hand oriented blades2.75" od.Assemble one or more impellers onto 7l drive shaft for mammalian cell culture.316l stainless steel constructionRa finish 32 micro inch or better. MADE IN THE USA
Catalog No. 50-194-6442 Supplier Bellco Glass, Inc. Supplier No. 790900033
Sst pitched blade impeller with right hand oriented blades2.75" od.Assemble one or more impellers onto 7l drive shaft for mammalian cell culture.316l stainless steel constructionRa finish 32 micro inch or better. MADE IN THE USA