Learn More
Bellco Glass, Inc. RH Pitch Blade Imp 3 1/2,15L
SDP
Catalog No. 501946488
Encompass_Preferred
Size: 15lMaterial: 316l sst with ra finish of 32 or betterDimensions: 3 1/2" x 2 3/16"Right hand pitch. MADE IN THE USA
Catalog No. 50-194-6488 Supplier Bellco Glass, Inc. Supplier No. 790915105
Size: 15lMaterial: 316l sst with ra finish of 32 or betterDimensions: 3 1/2" x 2 3/16"Right hand pitch. MADE IN THE USA