Learn More
Bellco Glass, Inc. RH Pitch Blade Impeller 2in,3L
SDP
Catalog No. 501946428
Encompass_Preferred
RH Pitch Blade ImpellerStainless Steel pitched blade impeller with right hand oriented blades.Assemble one or more impellers onto 3L 15L drive shaft for mammalian cell culture316L stainless steel construction. MADE IN THE USA
Catalog No. 50-194-6428 Supplier Bellco Glass, Inc. Supplier No. 790900016
RH Pitch Blade ImpellerStainless Steel pitched blade impeller with right hand oriented blades.Assemble one or more impellers onto 3L 15L drive shaft for mammalian cell culture316L stainless steel construction. MADE IN THE USA