Learn More
Abcam Recombinant human Ube2G1 protein (Active), 20UG
Supplier:  Abcam AB26910120UG
Encompass_Preferred
Recombinant human Ube2G1 protein (Active) is a Human Full Length protein, expressed in Escherichia coli, with >95% purity and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: UBE2G, UBE2G1, Ubiquitin-conjugating enzyme E2 G1, E2 ubiquitin-conjugating enzyme G1, E217K, UBC7, Ubiquitin carrier protein G1, Ubiquitin-protein ligase G1
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2153595