Learn More
Biotium RH237
Catalog No. 501964739
Encompass_Preferred
The styryl dye RH237 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons. Excitation/emission data (below)  is for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission.Properties: Ex/Em (MeOH) = 528/782 nm; Dark solid soluble in DMSO; Store at -20°C and protect from light; C29H40N2O3S; MW: 496.71
Catalog No. 50-196-4739 Supplier Biotium Supplier No. 61018
Add to Cart
Add to Cart
The styryl dye RH237 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons. Excitation/emission data (below)  is for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission.Properties: Ex/Em (MeOH) = 528/782 nm; Dark solid soluble in DMSO; Store at -20°C and protect from light; C29H40N2O3S; MW: 496.71