Learn More
Biotium RH421
Supplier:  Biotium 61017
Encompass_Preferred
RH421 is a fast responding membrane potential dye, exhibiting >20% fluorescence change per 100 mV on neuroblastoma cells.Properties: Ex/ Em = 515/704 nm (MeOH); Red solid soluble in DMSO, EtOH or DMF; Store at -20°C and protect from light, especially in solution; C29H42N2O3S; MW: 498.72; [107610-19-5]
Catalog No. 50-196-4738