Learn More
Biotium RH795
Catalog No. 501964740
Encompass_Preferred
The styryl dye RH795 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons. The dye is spectrally similar to RH414, but exhibits different physiological effects during staining. For example, RH414 causes arterial constriction during cortex staining, while RH795 does not. The excitation/emission data below are for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission. Properties: Ex/Em (MeOH) = 530/712 nm; Orange red solid soluble in H2O; Store at -20°C and protect from light; C26H39Br2N3O2; MW: 585.42; [172807-13-5]
Catalog No. 50-196-4740 Supplier Biotium Supplier No. 61019
The styryl dye RH795 is a fast-responding potentiometric probe that is primarily used for functional imaging of neurons. The dye is spectrally similar to RH414, but exhibits different physiological effects during staining. For example, RH414 causes arterial constriction during cortex staining, while RH795 does not. The excitation/emission data below are for the dye in methanol. In cell membranes, the spectra of styryl dyes are typically blue-shifted by as much as 20 nm for absorption or excitation and 80 nm for emission. Properties: Ex/Em (MeOH) = 530/712 nm; Orange red solid soluble in H2O; Store at -20°C and protect from light; C26H39Br2N3O2; MW: 585.42; [172807-13-5]