Learn More
Azzota Corp RHA-100T antifreeze refract
Supplier:  Azzota Corp RHA100T
Encompass_Preferred
RHA-100T Battery Fluid/Antifreeze/Cleaning Fluid Refractometer With ATC Ethylene Glycol -84 F to 32 F Propylene Glycol -60 F to 32 F Battery Fluid 1 100-1 400kg/l
Catalog No. 50-008-87305