Learn More
Azzota Corp RHA-800 antifreeze refract
Catalog No. 5000887284
Encompass_Preferred
RHA-800 Antifreeze Coolant Heavy-Duty Portable Refractometer for Battery/Antifreeze/Cleaning Fluid Ethylene Glycol -60 C to 0 C PG 0-100% BF 1 10-1 40KG/L with ATC
Catalog No. 50-008-87284 Supplier Azzota Corp Supplier No. RHA800
RHA-800 Antifreeze Coolant Heavy-Duty Portable Refractometer for Battery/Antifreeze/Cleaning Fluid Ethylene Glycol -60 C to 0 C PG 0-100% BF 1 10-1 40KG/L with ATC