Learn More
Medchemexpress LLC Rhamnetin | 90-19-7 | 10 MG
SDP
Catalog No. 5000418044
Encompass_Preferred
Rhamnetin is a quercetin derivative found in *Coriandrum sativum*. It functions as an inhibitor of secretory phospholipase A2 and histone deacetylase 2 (HDAC2). Rhamnetin has demonstrated antitumor, antioxidant, and anti-inflammatory activities.
  • Inhibits secretory phospholipase A2
  • Inhibits histone deacetylase 2 (HDAC2)
  • Demonstrates antitumor activity
  • Exhibits antioxidant properties
  • Provides anti-inflammatory effects
  • Inhibits proliferation of SKOV3 cancer cells
  • Arrests cell cycle at G1 phase
  • Induces apoptosis
  • Improves cognitive dysfunction in traumatic brain injury models
  • Inhibits inflammatory response and oxidative stress in the hippocampus
Catalog No. 50-004-18044 Supplier Medchemexpress LLC Supplier No. HYN703610MG
Rhamnetin is a quercetin derivative found in *Coriandrum sativum*. It functions as an inhibitor of secretory phospholipase A2 and histone deacetylase 2 (HDAC2). Rhamnetin has demonstrated antitumor, antioxidant, and anti-inflammatory activities.
  • Inhibits secretory phospholipase A2
  • Inhibits histone deacetylase 2 (HDAC2)
  • Demonstrates antitumor activity
  • Exhibits antioxidant properties
  • Provides anti-inflammatory effects
  • Inhibits proliferation of SKOV3 cancer cells
  • Arrests cell cycle at G1 phase
  • Induces apoptosis
  • Improves cognitive dysfunction in traumatic brain injury models
  • Inhibits inflammatory response and oxidative stress in the hippocampus