Learn More
Cayman Chemical Rhamnocitrin, Purity: ≥95%, CAS: 569-92-6, 10 mg
SDP
Catalog No. 502912376
Encompass_Preferred
A flavonoid with diverse biological activities; active against S. sonei, S. aureus, and M. luteus from 25-100 µg/ml; scavenges DPPH radicals in a cell-free assay (IC50 = ~25 µM); inhibits LPS- and IFN-γ-induced production of NO, TNF-α, and IL-6 in isolated mouse peritoneal macrophages
Catalog No. 50-291-2376 Supplier Cayman Chemical Supplier No. 3627210 MG
A flavonoid with diverse biological activities; active against S. sonei, S. aureus, and M. luteus from 25-100 µg/ml; scavenges DPPH radicals in a cell-free assay (IC50 = ~25 µM); inhibits LPS- and IFN-γ-induced production of NO, TNF-α, and IL-6 in isolated mouse peritoneal macrophages