Learn More
Cayman Chemical Rhamnopterin, Purity: ≥95%, CAS: 13392-24-0, 10 mg
SDP
Catalog No. 502913691
Encompass_Preferred
A derivative of biopterin; reduces the incidence of 4-dimethylaminoazobenzene-induced liver tumors in rats at 0.5% in the diet concurrently with 4-dimethylaminoazobenzene; has been used as an internal standard for the quantification of biopterin and neopterin in rat plasma by LC-MS
Catalog No. 50-291-3691 Supplier Cayman Chemical Supplier No. 3525410 MG
A derivative of biopterin; reduces the incidence of 4-dimethylaminoazobenzene-induced liver tumors in rats at 0.5% in the diet concurrently with 4-dimethylaminoazobenzene; has been used as an internal standard for the quantification of biopterin and neopterin in rat plasma by LC-MS