Learn More
Medchemexpress LLC HY-N1420 100mg Medchemexpress, Rhamnose CAS:3615-41-6 Purity:>98%
SDP
Catalog No. 501970335
Encompass_Preferred
Medchemexpress, HY-N1420 100mg Rhamnose CAS:3615-41-6 Rhamnose (L-Rhamnose) is a monosaccharide found in plants and bacteria. Rhamnose-conjugated immunogens is used in immunotherapies. Rhamnose crosses the epithelia via the transcellular pathway and acts as a marker of intestinal absorption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0335 Supplier Medchemexpress LLC Supplier No. HYN1420100MG
Medchemexpress, HY-N1420 100mg Rhamnose CAS:3615-41-6 Rhamnose (L-Rhamnose) is a monosaccharide found in plants and bacteria. Rhamnose-conjugated immunogens is used in immunotherapies. Rhamnose crosses the epithelia via the transcellular pathway and acts as a marker of intestinal absorption. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.