Learn More
Medchemexpress LLC Rhamnose monohydrate | 10030-85-0 | 182.17 | 25 MG
SDP
Catalog No. 5000389858
Encompass_Preferred
Rhamnose monohydrate (L-Rhamnose monohydrate) is an orally active deoxysugar. It can inhibit levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs) in skin aging models, and promote phosphorylation of PKA substrates and HSL in SVF-derived adipocytes, stimulating PKA signaling. It can also act against obesity in mice by stimulating fat dopamine receptors and inducing thermogenesis, and shows anti-aging effects.
  • Orally active deoxysugar
  • Inhibits levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs)
  • Stimulates PKA signaling
  • Induces thermogenesis and anti-obesity effects
  • Shows anti-aging effects
  • Used in the study of ehrlich's solid tumors and sarcomas
Catalog No. 50-003-89858 Supplier Medchemexpress LLC Supplier No. 25MGHYN1420AR
Rhamnose monohydrate (L-Rhamnose monohydrate) is an orally active deoxysugar. It can inhibit levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs) in skin aging models, and promote phosphorylation of PKA substrates and HSL in SVF-derived adipocytes, stimulating PKA signaling. It can also act against obesity in mice by stimulating fat dopamine receptors and inducing thermogenesis, and shows anti-aging effects.
  • Orally active deoxysugar
  • Inhibits levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs)
  • Stimulates PKA signaling
  • Induces thermogenesis and anti-obesity effects
  • Shows anti-aging effects
  • Used in the study of ehrlich's solid tumors and sarcomas