Learn More
Medchemexpress LLC Rhamnose (monohydrate) | 10030-85-0 | 182.17 | 250 G
SDP
Catalog No. 5000389524
Encompass_Preferred
Rhamnose monohydrate is an orally active deoxysugar. It can inhibit pro-inflammatory interleukin and matrix metalloproteinases in skin aging models. It also promotes PKA signaling, stimulating PKA substrates and HSL in SVF-derived adipocytes, and shows anti-aging effects. Additionally, it can act against obesity by stimulating fat dopamine receptors and inducing thermogenesis, and has been used in the study of Ehrlich's solid tumors and sarcomas.
  • Orally active deoxysugar.
  • Inhibits pro-inflammatory interleukin and matrix metalloproteinases (MMPs).
  • Promotes PKA signaling.
  • Acts against obesity.
  • Shows anti-aging effects.
  • Used in the study of Ehrlich's solid tumors and sarcomas.
Catalog No. 50-003-89524 Supplier Medchemexpress LLC Supplier No. 250GHYN1420A
Rhamnose monohydrate is an orally active deoxysugar. It can inhibit pro-inflammatory interleukin and matrix metalloproteinases in skin aging models. It also promotes PKA signaling, stimulating PKA substrates and HSL in SVF-derived adipocytes, and shows anti-aging effects. Additionally, it can act against obesity by stimulating fat dopamine receptors and inducing thermogenesis, and has been used in the study of Ehrlich's solid tumors and sarcomas.
  • Orally active deoxysugar.
  • Inhibits pro-inflammatory interleukin and matrix metalloproteinases (MMPs).
  • Promotes PKA signaling.
  • Acts against obesity.
  • Shows anti-aging effects.
  • Used in the study of Ehrlich's solid tumors and sarcomas.