Learn More
Medchemexpress LLC Rhamnose monohydrate | 10030-85-0 | 182.17 | 50 MG
SDP
Catalog No. 5000414850
Encompass_Preferred
Rhamnose monohydrate is an orally active deoxysugar and an analytical standard for Rhamnose monohydrate. It inhibits pro-inflammatory interleukin and matrix metalloproteinases in skin aging models and stimulates PKA signaling in SVF-derived adipocytes. This compound also exhibits anti-obesity effects by stimulating fat dopamine receptors and inducing thermogenesis, and has shown anti-aging and anti-tumor effects in studies involving Ehrlich's solid tumors and sarcomas.
  • Inhibits pro-inflammatory interleukin and matrix metalloproteinases in skin aging models.
  • Promotes phosphorylation levels of PKA substrates and HSL.
  • Acts against obesity by stimulating fat dopamine receptors and inducing thermogenesis.
  • Shows anti-aging effects.
  • Can be used in the study of Ehrlich's solid tumors and sarcomas.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments.
Catalog No. 50-004-14850 Supplier Medchemexpress LLC Supplier No. HYN1420AR50MG
Rhamnose monohydrate is an orally active deoxysugar and an analytical standard for Rhamnose monohydrate. It inhibits pro-inflammatory interleukin and matrix metalloproteinases in skin aging models and stimulates PKA signaling in SVF-derived adipocytes. This compound also exhibits anti-obesity effects by stimulating fat dopamine receptors and inducing thermogenesis, and has shown anti-aging and anti-tumor effects in studies involving Ehrlich's solid tumors and sarcomas.
  • Inhibits pro-inflammatory interleukin and matrix metalloproteinases in skin aging models.
  • Promotes phosphorylation levels of PKA substrates and HSL.
  • Acts against obesity by stimulating fat dopamine receptors and inducing thermogenesis.
  • Shows anti-aging effects.
  • Can be used in the study of Ehrlich's solid tumors and sarcomas.
  • Used as an analytical standard in qualitative, quantitative, and methodological research experiments.