Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rhamnose monohydrate | 10030-85-0 | 182.17 | 500 G
SDP

Catalog No. 5000415978
Encompass_Preferred

Rhamnose monohydrate (L-Rhamnose monohydrate) is an orally active deoxysugar. It can inhibit levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs) in skin aging models. It promotes the phosphorylation levels of PKA substrates and HSL in SVF-derived adipocytes, stimulating PKA signaling. Rhamnose monohydrate can act against obesity in mice by stimulating fat dopamine receptors and inducing thermogenesis and shows anti-aging effects. It can be used in the study of Ehrlich's solid tumors and sarcomas.

  • Purity: 99.95%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Plants other families, microorganisms, widespread

Catalog No. 50-004-15978 Supplier Medchemexpress LLC Supplier No. HYN1420A500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rhamnose monohydrate (L-Rhamnose monohydrate) is an orally active deoxysugar. It can inhibit levels of pro-inflammatory interleukin and matrix metalloproteinases (MMPs) in skin aging models. It promotes the phosphorylation levels of PKA substrates and HSL in SVF-derived adipocytes, stimulating PKA signaling. Rhamnose monohydrate can act against obesity in mice by stimulating fat dopamine receptors and inducing thermogenesis and shows anti-aging effects. It can be used in the study of Ehrlich's solid tumors and sarcomas.

  • Purity: 99.95%
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Saccharides, monosaccharides
  • Initial source: Plants other families, microorganisms, widespread

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.