Learn More
Medchemexpress LLC Rhamnose monohydrate | 10030-85-0 | MFCD00149363 | 182.17 | 50 G
SDP
Catalog No. 5000414534
Encompass_Preferred
Rhamnose monohydrate (L-Rhamnose monohydrate) is an orally active deoxysugar with diverse biological activities. It is capable of inhibiting pro-inflammatory interleukin and matrix metalloproteinases in skin aging models.
  • Promotes phosphorylation levels of PKA substrates and HSL in SVF-derived adipocytes.
  • Stimulates PKA signaling.
  • Shows anti-obesity effects by stimulating fat dopamine receptors and inducing thermogenesis in mice.
  • Exhibits anti-aging effects.
  • Can be utilized in the study of Ehrlich's solid tumors and sarcomas.
Catalog No. 50-004-14534 Supplier Medchemexpress LLC Supplier No. HYN1420A50G
Rhamnose monohydrate (L-Rhamnose monohydrate) is an orally active deoxysugar with diverse biological activities. It is capable of inhibiting pro-inflammatory interleukin and matrix metalloproteinases in skin aging models.
  • Promotes phosphorylation levels of PKA substrates and HSL in SVF-derived adipocytes.
  • Stimulates PKA signaling.
  • Shows anti-obesity effects by stimulating fat dopamine receptors and inducing thermogenesis in mice.
  • Exhibits anti-aging effects.
  • Can be utilized in the study of Ehrlich's solid tumors and sarcomas.