Learn More
TARGETMOL CHEMICALS INC RHAPONTIGENIN 10MG
SDP
Catalog No. 502580126
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rhapontigenin (Protigenin) is a mechanism-based selective inactivator of cytochrome P450 1A1 (IC50 400 nM) an aryl hydrocarbon hydroxylase which activates polycyclic aromatic hydrocarbons that act as procarcinogens. Rhapontigenin is a natural analog of resveratrol with antioxidant and anti-cancer activity. At higher concentrations rhapontigenin inhibits the proliferation of HL-60R and HepG2 cancer cell lines (IC50 48 uM). purity: 99%
Catalog No. 50-258-0126 Supplier TARGETMOL CHEMICALS INC Supplier No. T377610MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Rhapontigenin (Protigenin) is a mechanism-based selective inactivator of cytochrome P450 1A1 (IC50 400 nM) an aryl hydrocarbon hydroxylase which activates polycyclic aromatic hydrocarbons that act as procarcinogens. Rhapontigenin is a natural analog of resveratrol with antioxidant and anti-cancer activity. At higher concentrations rhapontigenin inhibits the proliferation of HL-60R and HepG2 cancer cell lines (IC50 48 uM). purity: 99%