Learn More
Medchemexpress LLC Rhapontigenin | 500-65-2 | MFCD00017718 | ">98.0%" | 10 MG
SDP
98.0%" | 10 MG">
Catalog No. 5000296026
Encompass_Preferred
Rhapontigenin is a naturally occurring stilbenoid used as a research-grade bioactive compound and analytical standard; it is reported to inactivate cytochrome P450 1A1 and shows antioxidant and anticancer activities.
  • High purity (≥98%) suitable for research applications.
  • Solid powder form, soluble in common organic solvents such as DMSO.
  • Stable when stored under recommended conditions: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Used in enzymology, cell-based assays, and as an analytical reference standard.
Catalog No. 50-002-96026 Supplier Medchemexpress LLC Supplier No. HYN222910MG
Rhapontigenin is a naturally occurring stilbenoid used as a research-grade bioactive compound and analytical standard; it is reported to inactivate cytochrome P450 1A1 and shows antioxidant and anticancer activities.
  • High purity (≥98%) suitable for research applications.
  • Solid powder form, soluble in common organic solvents such as DMSO.
  • Stable when stored under recommended conditions: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Used in enzymology, cell-based assays, and as an analytical reference standard.