Learn More
BD Rhapsody™ ATAC-Seq Amplification Kit
Catalog No. BDB940519 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
ATAC-Seq Amplification Kit
Catalog No. Product Type
BDB940519 ATAC-Seq Amplification Kit
1 options
Catalog No. BDB940519 Supplier BD Supplier No. 571356
Add to Cart
Add to Cart
The BD Rhapsody™ ATAC-Seq Assays enable you to either generate highly reproducible open chromatin profiling data from single cells or perform a multiomic analysis of open chromatin accessibility and transcriptome of single cells in one experiment using the BD Rhapsody™ Single-Cell Analysis System.
Validated on the new BD Rhapsody™ Enhanced Cell Capture Beads V3, BD Rhapsody™ ATAC-Seq Assays generate consistent, high-quality data across different samples and users and a wide range of cell inputs. Key features include:
  • High sensitivity and specificity across different experimental conditions
  • Capable of accommodating a wide range of cell inputs
  • Integrated epigenomic and transcriptomic characterization on the same cells
  • Compatibility with Custom BD™ Nuclear Antibody-Oligonucleotide Conjugates
TRUSTED_SUSTAINABILITY
Specifications
Product Type ATAC-Seq Amplification Kit