Description
Validated on the new BD Rhapsody™ Enhanced Cell Capture Beads V3, BD Rhapsody™ ATAC-Seq Assays generate consistent, high-quality data across different samples and users and a wide range of cell inputs. Key features include:
- High sensitivity and specificity across different experimental conditions
- Capable of accommodating a wide range of cell inputs
- Integrated epigenomic and transcriptomic characterization on the same cells
- Compatibility with Custom BD™ Nuclear Antibody-Oligonucleotide Conjugates
Specifications
Specifications
| Product Type | ATAC-Seq Tagmentation and Supplemental Reagents Kit |