Learn More
BD Rhapsody™ cDNA Kit
Catalog No. BDB633773 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BDB633773 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. BDB633773 Supplier BD Supplier No. 633773
Add to Cart
Add to Cart
This item is not returnable. View return policy
The kit can be used with the BD Rhapsody™ Targeted Assay, BD Rhapsody™ Whole Transcriptome Analysis Amplification Kit, BD Rhapsody™ TCR/BCR Profiling Assays, BD™ Single-Cell Multiplexing Kit, and BD™ AbSeq Assays.
  • Once the RNA and protein information are captured from single cells on the BD Rhapsody™ Cell Capture Beads, reagents provided in the BD Rhapsody™ cDNA Kit allow you to perform reverse transcription and generate cDNA for use as a template for all your desired downstream library preparations
  • The kit can be used with the BD Rhapsody™ Targeted Assay, BD Rhapsody™ Whole Transcriptome Analysis Amplification Kit, BD Rhapsody™ TCR/BCR Profiling Assays, BD™ Single-Cell Multiplexing Kit and BD™ AbSeq Assays
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.