Learn More
BD Rhapsody™ Enhanced Cartridge Reagent Kit V3
Catalog No. BDB940515 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
Enhanced Cartridge Reagent Kit V3
Catalog No. Product Type
BDB940515 Enhanced Cartridge Reagent Kit V3
1 options
Catalog No. BDB940515 Supplier BD Supplier No. 667052
Add to Cart
Add to Cart
This kit provides the necessary reagents to support the amplification of single cell libraries using the BD Rhapsody™ Single-Cell Analysis system
The purpose of this kit is to provide the necessary reagents needed to prepare the BD Rhapsody™ Cartridges, extract RNA and antibody oligos bound to the cells, and capture them on BD Rhapsody™ Enhanced Cell Capture Beads.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Enhanced Cartridge Reagent Kit V3