Learn More
BD Rhapsody™ Express
Catalog No. BDB633702 Shop All BD Biosciences Products
Change view
Click to view available options
BD Rhapsody
Catalog No.
BDB633702 BD Rhapsody
1 options
This item is not returnable. View return policy
Catalog No. BDB633702 Supplier BD Supplier No. 633702
This item is not returnable. View return policy
The BD Rhapsody™ Express Single-Cell Analysis System is ideal for routine use with familiar cell systems that do not require visual QC or troubleshooting.
  • Enables single-cell capture and barcoding of hundreds to thousands of single cells for analysis of genomic and proteomic information, using proprietary, gentle, robust microwell-based single-cell partitioning technology
  • Ideal for routine use with familiar cell systems that do not require visual QC or troubleshooting
TRUSTED_SUSTAINABILITY
Specifications
Product Line BD Rhapsody