Description
The BD Rhapsody™ HT Xpress System offers the same performance as the BD Rhapsody™ Express System, now featuring a new flexible cartridge design with the ability to run up to eight times the number of lanes at one time with a broad range of cell input, from 100–800,000 cells per cartridge.
Expanded Throughput
- Million-cell studies now possible
- Capable of capturing more than 640,000 cells per cartridge
- Up to 80% cell capture rate (for certain cell types)
- Low multiplet rate per lane
Flexible Cartridge Design
- Up to 192 samples per cartridge (up to 24 samples per lane)
- Partial use of cartridge enables:
- Running more or different types of experiments
Maintain Sample Integrity
- Gentle microwell technology
- No sample loss due to clogging of channels
- Recover cells with disparate size and morphology, including fragile cell types
- Subsample and archive cell capture beads for flexibility with experimental design and collaborating across sites with equivalent data obtained from fresh beads
Specifications
Specifications
| Product Line | BD Rhapsody |