Learn More
BD Rhapsody™ Immune Response Panel HS
Catalog No. BDB633750 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
4 Tests
Catalog No. Quantity
BDB633750 4 Tests
1 options
This item is not returnable. View return policy
Catalog No. BDB633750 Supplier BD Supplier No. 633750
Add to Cart
Add to Cart
This item is not returnable. View return policy
This panel contains primer pairs that target 399 genes commonly expressed in human immune cells.
  • The primer pairs have been validated for use in BD Rhapsody™ targeted assays
TRUSTED_SUSTAINABILITY
Specifications
No. of Tests 4
Quantity 4 Tests