Learn More
BD Rhapsody™ Immune Response Panel Mm
Catalog No. BDB633753 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
4 Tests
Catalog No. Quantity
BDB633753 4 Tests
1 options
This item is not returnable. View return policy
Catalog No. BDB633753 Supplier BD Supplier No. 633753
Add to Cart
Add to Cart
This item is not returnable. View return policy
This panel contains primer pairs that target 397 genes commonly found in mouse immune cells.
  • The primer pairs have been validated for use in BD Rhapsody™ targeted assays
TRUSTED_SUSTAINABILITY
Specifications
No. of Tests 4
Quantity 4 Tests