Learn More
BD Rhapsody™ Onco-BC Panel HS
Catalog No. BDB633752 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
4 Tests
Catalog No. Quantity
BDB633752 4 Tests
1 options
This item is not returnable. View return policy
Catalog No. BDB633752 Supplier BD Supplier No. 633752
Add to Cart
Add to Cart
This item is not returnable. View return policy
This panel contains primer pairs that target 389 genes commonly expressed in human breast cancer samples that contain breast cancer cells, tumor infiltrating immune cells, and other cell types present in tumor microenvironments.
  • The primer pairs have been validated for use in BD Rhapsody™ targeted assays
TRUSTED_SUSTAINABILITY
Specifications
No. of Tests 4
Quantity 4 Tests