Learn More
BD Rhapsody™ Targeted mRNA and AbSeq Amplification Kit
Catalog No. BDB633774 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
BDB633774 1 Ea.
1 options
This item is not returnable. View return policy
Catalog No. BDB633774 Supplier BD Supplier No. 633774
Add to Cart
Add to Cart
This item is not returnable. View return policy
The purpose of this kit is to provide the necessary reagents for the preparation of single cell mRNA, Sample Tag, and BD™ AbSeq libraries with the BD Rhapsody™ Single-Cell Analysis system or the BD Rhapsody™ Express Single-Cell Analysis system.
  • Provides the necessary reagents for the preparation of single cell mRNA, Sample Tag, and BD™ AbSeq libraries with the BD Rhapsody™ Single-Cell Analysis system or the BD Rhapsody™ Express Single-Cell Analysis system
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.