Learn More
BD Rhapsody™ TCR/BCR Next Amplification Kit
Catalog No. BDB940514 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
TCR/BCR Next Amplification Kit
Catalog No. Product Type
BDB940514 TCR/BCR Next Amplification Kit
1 options
Catalog No. BDB940514 Supplier BD Supplier No. 667058
Add to Cart
Add to Cart
The next generation of BD Rhapsody™ TCR/BCR Multiomic Assays reveal immune repertoire richness with great sensitivity alongside simultaneous single-cell gene and protein expression analyses.
Validated on our new Enhanced Cell Capture Beads V3, BD Rhapsody™ TCR/BCR Next Multiomic Assays enable a highly sensitive analysis of full-length TCR/BCR repertoires with robust workflows for human and mouse. Key features include:
  • Superior full-length TCR and BCR pairing metrics
  • Multiomics-enabled for simultaneous immune profiling alongside protein and gene expression analyses in the same cell
  • Compatibility with preserved cells and BD™ OMICS-Guard Sample Preservation Buffer
  • Intuitive data analysis pipeline that provides unbiased insights about the biology of your samples
TRUSTED_SUSTAINABILITY
Specifications
Product Type TCR/BCR Next Amplification Kit