Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Rhapsody™ TCR/BCR Next Multiomic Assay for WTA

Catalog No. BDB667401 Shop All BD Biosciences Products
Change view
Click to view available options
No. of Samples:
8 Samples
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Samples
BDB667401 8 Samples
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB667401 Supplier BD Supplier No. 667400
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The next generation of BD Rhapsody™ TCR/BCR Multiomic Assays reveal immune repertoire richness with great sensitivity alongside simultaneous single-cell gene and protein expression analyses.

Validated on our new Enhanced Cell Capture Beads V3, BD Rhapsody™ TCR/BCR Next Multiomic Assays enable a highly sensitive analysis of full-length TCR/BCR repertoires with robust workflows for human and mouse. Key features include:

  • Superior full-length TCR and BCR pairing metrics
  • Multiomics-enabled for simultaneous immune profiling alongside protein and gene expression analyses in the same cell
  • Compatibility with preserved cells and BD™ OMICS-Guard Sample Preservation Buffer
  • Intuitive data analysis pipeline that provides unbiased insights about the biology of your samples
TRUSTED_SUSTAINABILITY

Specifications

Species Human
No. of Samples 8 Samples
Target Total RNA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.