Learn More
BD™ Rhapsody™ Whole Transcriptome Analysis (WTA) Reagent Kit
Catalog No. BDB665915 Shop All BD Biosciences Products
Change view
Click to view available options
BD™ Whole Transcriptome Analysis
Catalog No.
BDB665915 BD™ Whole Transcriptome Analysis
1 options
Catalog No. BDB665915 Supplier BD™ Supplier No. 665915
The BD Rhapsody™ Whole Transcriptome Analysis (WTA) Amplification kit enables unbiased three prime end capture, amplification and detection of transcriptomes at a single cell level.
The BD Rhapsody WTA Amplification kit has been thoroughly tested and validated to generate consistent, high-quality whole transcriptome data across different workflows, different users, and a wide range of cell inputs.
  • Identify genes of interest that can be used for building your targeted panels.
  • Customizable targeted panel creation makes it easy to focus on the genes that really matter to your research.
  • With the BD Single Cell Multiplexing kit (SMK) and the Rhapsody WTA Amplification kit working seamlessly together, you can save time and money by running multiple samples at once, giving you the option of multiplexing without compromising the quality of your data.
  • Uncover the whole transcriptome of a single cell is at your fingertips.
  • This catalog number covers all the 4 kits needed for your single cell WTA experiment and includes:
    • BD Rhapsody™ Enhanced Cartridge Reagent Kit (cat no. 664887)
    • BD Rhapsody™ Cartridge Kit (cat no. 633733)
    • BD Rhapsody™ cDNA Kit (cat no. 633773)
    • BD Rhapsody™ Whole Transcriptome Analysis (WTA) Amplification Kit (cat no. 633801)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) BD Rhapsody Platform, Single-Cell Analysis System
Product Type Whole Transcriptome