Learn More
Azzota Corp RHB-18T aluminum Brix refract
Catalog No. 5000887308
Encompass_Preferred
RHB-18T Heavy-Duty Portable Refractometer Sturdy design made of Aluminum Brix Range 0-18% Brix Min Div 1% Brix Accuracy /-1 0% With ATC
Catalog No. 50-008-87308 Supplier Azzota Corp Supplier No. RHB18T
RHB-18T Heavy-Duty Portable Refractometer Sturdy design made of Aluminum Brix Range 0-18% Brix Min Div 1% Brix Accuracy /-1 0% With ATC