Learn More
Takara Bio RHB-A, 500 mL
SDP
Catalog No. NC0978847
Encompass
Maintenance and expansion of adherent, mouse and human neural stem (NS) cells. Neural differentiation of mouse embryonic stem (ES) cells in monolayer culture. Derivation of NS cells from mouse ES cells and fetal/adult tissues. Differentiation of mouse and human NS cells into functional neurons. Maintenance and expansion of human glioblastoma stem cell lines.
Catalog No. NC0978847 Supplier Takara Bio Supplier No. Y40001
Maintenance and expansion of adherent, mouse and human neural stem (NS) cells. Neural differentiation of mouse embryonic stem (ES) cells in monolayer culture. Derivation of NS cells from mouse ES cells and fetal/adult tissues. Differentiation of mouse and human NS cells into functional neurons. Maintenance and expansion of human glioblastoma stem cell lines.